Learn More
Iridium(IV) iodide, Premion™, 99.95% (metals basis), Ir 27.0% min, Thermo Scientific Chemicals
Description
Methanol carbonylation to acetic acid is catalysed with high rates at low water concentrations using an iridium/iodide based catalyst. Iridium(IV)-iodide mixtures participates in the Investigations in platinum metal group electrochemistry. Rhodium- and iridium-iodide catalysts also promote the carbonylation of higher alcohols to linear or branched chain carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Insoluble in water and alcohol; Soluble in aqueous KI |
Formula Weight | 699.82 |
Percent Purity | 99.95% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Ir 27.0% min. |
Assay | (metals basis) |
Chemical Name or Material | Iridium(IV) iodide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.