missing translation for 'onlineSavingsMsg'
Learn More

Iridium(IV) chloride, 99.95% (metals basis), Ir 56.5 min, Thermo Scientific Chemicals

Product Code. 11376568
Click to view available options
Quantity:
0.25 g
5 g

Iridium(IV) chloride is used in chemical analysis as testing for nitric acid in the presence of nitrous acid, microscopy and in plating solutions. It acts as a catalyst precursor for the stereoselective reduction of cyclohexanones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-97-5
Molecular Formula Cl4Ir
Molecular Weight (g/mol) 334.017
MDL Number MFCD00016155
InChI Key CALMYRPSSNRCFD-UHFFFAOYSA-J
Synonym iridium iv chloride, iridium tetrachloride, iridium chloride ircl4, iridic chloride, unii-pcg7kvc21i, pcg7kvc21i, acmc-1cp8w, iridium iv chloride, premion
PubChem CID 24815
IUPAC Name tetrachloroiridium
SMILES Cl[Ir](Cl)(Cl)Cl

Specifications

Melting Point (decomposition)
Physical Form Glassy Amorphous Pieces, Powder
Quantity 5 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Soluble in water,alcohol and dilute hydrochloric acid.
Formula Weight 334.01
Sensitivity Hygroscopic
Chemical Name or Material Iridium(IV) chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'