missing translation for 'onlineSavingsMsg'
Learn More

(Iodomethyl)trimethylsilane, 99%, Thermo Scientific Chemicals

77.79 € - 298.49 €

Chemical Identifiers

CAS 4206-67-1
Molecular Formula C4H11ISi
Molecular Weight (g/mol) 214.121
MDL Number MFCD00001077
InChI Key VZNYXGQMDSRJAL-UHFFFAOYSA-N
Synonym iodomethyl trimethylsilane, iodomethyl-trimethylsilane, iodomethyl trimethyl silane, trimethylsilyl methyl iodide, sila-neopentyliodide, trimethylsilylmethyl iodide, acmc-209jn7, trimethyl iodomethyl silane, iodanylmethyl trimethyl silane
PubChem CID 77877
IUPAC Name iodomethyl(trimethyl)silane
SMILES C[Si](C)(C)CI
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11451960
View Documents
Thermo Scientific Alfa Aesar
A15120.09
10 g
77.79 €
10g
Please to purchase this item. Need a web account? Register with us today!
11461960
View Documents
Thermo Scientific Alfa Aesar
A15120.18
50 g
298.49 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

(Iodomethyl)trimethylsilane is used for the N-alkylation of amides, in a reaction sequence to prepare unstabilized ylides for [2+3]-cycloadditions. It can react with acetoacetic acid ethyl ester to get 3-trimethylsilanyl-proπonic acid ethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Iodomethyl)trimethylsilane is used for the N-alkylation of amides, in a reaction sequence to prepare unstabilized ylides for [2+3]-cycloadditions. It can react with acetoacetic acid ethyl ester to get 3-trimethylsilanyl-proπonic acid ethyl ester.

Solubility
Difficult to mix.

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Chemical Identifiers

4206-67-1
214.121
VZNYXGQMDSRJAL-UHFFFAOYSA-N
77877
C[Si](C)(C)CI
C4H11ISi
MFCD00001077
iodomethyl trimethylsilane, iodomethyl-trimethylsilane, iodomethyl trimethyl silane, trimethylsilyl methyl iodide, sila-neopentyliodide, trimethylsilylmethyl iodide, acmc-209jn7, trimethyl iodomethyl silane, iodanylmethyl trimethyl silane
iodomethyl(trimethyl)silane

Specifications

140°C to 142°C
C4H11ISi
MFCD00001077
1731485
iodomethyl trimethylsilane, iodomethyl-trimethylsilane, iodomethyl trimethyl silane, trimethylsilyl methyl iodide, sila-neopentyliodide, trimethylsilylmethyl iodide, acmc-209jn7, trimethyl iodomethyl silane, iodanylmethyl trimethyl silane
VZNYXGQMDSRJAL-UHFFFAOYSA-N
iodomethyl(trimethyl)silane
77877
99%
1.4915
1.443
4206-67-1
(CH3)3SiCH2I
UN1993
31°C (88°F)
Difficult to mix.
C[Si](C)(C)CI
214.121
214.12
Recognizable
Light sensitive
(Iodomethyl)trimethylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 224-123-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.