missing translation for 'onlineSavingsMsg'
Learn More

Iodocyclohexane, 98%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11446626
Click to view available options
Quantity:
25 g
100 g
500 g

Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodocyclohexane has been employed as reagent in demethylation of aryl methyl ethers in DMF under reflux condition.

Solubility
Soluble in ethanol, ether, acetone.

Notes
Light sensitive. Store away from oxidizing agent, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-62-0
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD00003826
InChI Key FUCOMWZKWIEKRK-UHFFFAOYSA-N
Synonym cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i
PubChem CID 12289
IUPAC Name iodocyclohexane
SMILES IC1CCCCC1

Specifications

Density 1.625
Boiling Point 180°C
Flash Point 71°C (159°F)
Refractive Index 1.5475
Quantity 100 g
Beilstein 1900797
Sensitivity Light sensitive
Solubility Information Soluble in ethanol,ether,acetone.
Formula Weight 210.06
Percent Purity 98%
Chemical Name or Material Iodocyclohexane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.