Learn More
Iodoacetyl chloride, 97%, Thermo Scientific Chemicals
108.54 € - 426.12 €
Chemical Identifiers
CAS | 38020-81-4 |
---|---|
Molecular Formula | C2H2ClIO |
Molecular Weight (g/mol) | 204.39 |
MDL Number | MFCD00013718 |
InChI Key | BSVMPWANOMFSPR-UHFFFAOYSA-N |
Synonym | iodoacetyl chloride, iodoacetylchloride, acetyl chloride, iodo, jodacetylchlorid, acetylchloride,iodo, iodoacetic acid chloride, acmc-1aewu, iodoacetyl chloride 5g, acetyl chloride, iodo-6ci,9ci |
PubChem CID | 3084680 |
IUPAC Name | 2-iodoacetyl chloride |
SMILES | ClC(=O)CI |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351577
|
Thermo Scientific Alfa Aesar
L04428.06 |
5 g |
108.54 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361577
|
Thermo Scientific Alfa Aesar
L04428.14 |
25 g |
426.12 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Iodoacetyl chloride is used for biochemical research purposes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIodoacetyl chloride is used for biochemical research purposes.
Solubility
Miscible with water.
Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong reducing agents. Reacts with water.
Chemical Identifiers
38020-81-4 | |
204.39 | |
BSVMPWANOMFSPR-UHFFFAOYSA-N | |
3084680 | |
ClC(=O)CI |
C2H2ClIO | |
MFCD00013718 | |
iodoacetyl chloride, iodoacetylchloride, acetyl chloride, iodo, jodacetylchlorid, acetylchloride,iodo, iodoacetic acid chloride, acmc-1aewu, iodoacetyl chloride 5g, acetyl chloride, iodo-6ci,9ci | |
2-iodoacetyl chloride |
Specifications
38020-81-4 | |
48°C to 50°C (12 mmHg) | |
MFCD00013718 | |
1739078 | |
iodoacetyl chloride, iodoacetylchloride, acetyl chloride, iodo, jodacetylchlorid, acetylchloride,iodo, iodoacetic acid chloride, acmc-1aewu, iodoacetyl chloride 5g, acetyl chloride, iodo-6ci,9ci | |
BSVMPWANOMFSPR-UHFFFAOYSA-N | |
2-iodoacetyl chloride | |
3084680 | |
97% |
2.28 | |
C2H2ClIO | |
UN3265 | |
Light and moisture sensitive | |
Miscible with water. | |
ClC(=O)CI | |
204.39 | |
204.39 | |
Iodoacetyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 253-748-7
missing translation for 'tsca' : No
Recommended Storage : Keep cold
- Iodoacetyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Iodoacetyl chloride
RUO – Research Use Only