Learn More
Iodoacetamide, 98%, Thermo Scientific Chemicals
31.56 € - 1270.32 €
Chemical Identifiers
CAS | 144-48-9 |
---|---|
Molecular Formula | C2H4INO |
Molecular Weight (g/mol) | 184.96 |
InChI Key | PGLTVOMIXTUURA-UHFFFAOYSA-N |
Synonym | iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid |
PubChem CID | 3727 |
IUPAC Name | 2-iodoacetamide |
SMILES | C(C(=O)N)I |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10346660
|
Thermo Scientific Acros
122270050 |
5 g |
31.56 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10408660
|
Thermo Scientific Acros
122270250 |
25 g |
108.54 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10235260
|
Thermo Scientific Acros
122271000 |
100 g |
279.39 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11955461
|
Thermo Scientific Acros
122275000 |
500 g |
1270.32 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
144-48-9 | |
184.96 | |
iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid | |
2-iodoacetamide |
C2H4INO | |
PGLTVOMIXTUURA-UHFFFAOYSA-N | |
3727 | |
C(C(=O)N)I |
Specifications
144-48-9 | |
Beige or White to Yellow | |
97.5% min. (ex I) (Argentometry) | |
C2H4INO | |
02,223 | |
iodoacetamide, monoiodoacetamide, surauto, acetamide, 2-iodo, 2-iodo-acetamide, usaf d-1, alpha-iodoacetamide, ccris 7710, unii-zrh8m27s79, n-3-methoxy-4-butoxybenzyl thiobarbituric acid | |
PGLTVOMIXTUURA-UHFFFAOYSA-N | |
2-iodoacetamide | |
3727 | |
98% | |
Iodoacetamide |
>90.0°C | |
Authentic | |
Glass bottle | |
ICH2CONH2 | |
01,504 | |
Solubility in water: slightly soluble | |
C(C(=O)N)I | |
184.96 | |
184.96 | |
Crystals or Powder |
Safety and Handling
GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Causes severe skin burns and eye damage.
Toxic if swallowed.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-630-1
- Iodoacetamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only