missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine solution, 0.01M EU Pharmacopoeia, Fisher Chemical™
Click to view available options
Quantity:
1000 mL
Packaging:
Glass Bottle
Specifications
Specifications
Concentration | 0.01M |
CAS | 7732-18-5 |
Packaging | Glass Bottle |
Quantity | 1000 mL |
Grade | Eu. Ph. Reference |
Chemical Name or Material | Iodine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction