missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine Solution, 0.05 M, Honeywell Fluka™
Volumetric, 0.05 M I2 (0.1N)
48.81 € - 51.73 €
Chemical Identifiers
CAS | 7553-56-2 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
SMILES | II |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15633540
|
Honeywell-Fluka
318981-100ML |
100 mL |
51.73 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15643540
|
Honeywell-Fluka
318981-500ML |
500 mL |
48.81 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
II |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 |
Specifications
7553-56-2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
0.05 M | |
Iodine Solution |
I2 | |
3587194 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
diiodine | |
807 | |
253.81g/mol | |
1.034g/mL at 25°C |
Safety and Handling
H373
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Iodine solution
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : iodine
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Iodine Solution, 0.05 M, Honeywell Fluka™