missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, Reagent Grade, chips, ≥99%, Honeywell™
Reagent Grade, chips,≥99%
49.09 € - 3473.15 €
Chemical Identifiers
CAS | 7553-56-2 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
SMILES | II |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15675150
|
Honeywell Research Chemicals
376558-50G |
50 g | Glass Bottle | ||||||
15665150
|
Honeywell Research Chemicals
376558-500G |
500 g | Glass Bottle | ||||||
15655150
|
Honeywell Research Chemicals
376558-2.5KG |
2.5 kg | Glass Bottle | ||||||
15645150
|
Honeywell Research Chemicals
376558-12KG |
12 kg | Drum With Inliner | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7553-56-2 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
II |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 |
Specifications
184°C (literature) | |
7553-56-2 | |
I2 | |
MFCD00011355 MFCD00164163 | |
3587194 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
molecular iodine | |
807 | |
253.81 | |
Reagent | |
Iodine, Reagent Grade, chips, ≥99% |
113°C (literature) | |
100.0000% | |
CHCl3 | |
UN3495 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
≥99% | |
4.930 g/cm3 |
Safety and Handling
missing translation for 'shelfLife' : 1620 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Iodine
missing translation for 'einecsNumber' : 231-442-4
Recommended Storage : Room Temp