missing translation for 'onlineSavingsMsg'
Learn More

Iodine monobromide, 98%, Thermo Scientific Chemicals

32.76 € - 180.90 €

Chemical Identifiers

CAS 7789-33-5
Molecular Formula BrI
Molecular Weight (g/mol) 206.81
MDL Number MFCD00011353
InChI Key CBEQRNSPHCCXSH-UHFFFAOYSA-N
Synonym iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
PubChem CID 82238
IUPAC Name iodobromane
SMILES BrI
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11428880
SDS View Documents
Thermo Scientific Alfa Aesar
A16158.14
25 g
32.76 €
25g
Please to purchase this item. Need a web account? Register with us today!
11438880
SDS View Documents
Thermo Scientific Alfa Aesar
A16158.22
100 g
92.76 €
100g
Please to purchase this item. Need a web account? Register with us today!
11443217
SDS View Documents
Thermo Scientific Alfa Aesar
A16158.30
250 g
180.90 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodine monobromide is used as an electrophile employed in a new synthetic approach to polyketide structural units. It is also used in iodometry and serves as a source of I+. It is a powerful iodinating agent used in organic synthesis as well as involved in the synthesis of radioiodinated fatty acids for heart imaging.

Solubility
Soluble in water, alcohol, ether, carbon disulfide and glacial acetic acid.

Notes
Air, light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with alcohols, phosphorus, sodium and sodiumoxides and potassium.
Specifications

Chemical Identifiers

7789-33-5
206.81
CBEQRNSPHCCXSH-UHFFFAOYSA-N
82238
BrI
BrI
MFCD00011353
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
iodobromane

Specifications

116°C (decomposition)
7789-33-5
MFCD00011353
3902973
14,5016
Soluble in water,alcohol,ether,carbon disulfide and glacial acetic acid.
BrI
206.81
206.81
Pungent
4.416
50°C (sublimation)
BrI
UN3260
None
iodine monobromide, iodine bromide, iodobromine, iodine bromide ibr, unii-g0k622rd9n, iodinemonobromide, iodobromane, bromoiodine, jodmonobromid, hanus solution
CBEQRNSPHCCXSH-UHFFFAOYSA-N
iodobromane
82238
98%
Light and moisture sensitive
Iodine monobromide
Video's
Veiligheid en behandeling

Veiligheid en behandeling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 232-159-9

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold; Store under Nitrogen

Productidentificatie
  • Iodine monobromide

Signaalwoord
  • Danger

Gevarencategorie
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Gevarenaanduiding
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.

Veiligheidsmaatregel
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Aanvullende informatie
  • MIXTURE LIST-Contains : Iodine bromide (IBr)

SDS
Documentatie

Documentatie

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine monobromide, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.