missing translation for 'onlineSavingsMsg'
Learn More

Iodine lump, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 15402057
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Iodine lump is used in OLED materials, pharmaceuticals, and the electronics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7553-56-2
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

Melting Point 183°C to 186°C (sublimation)
Physical Form Solid
Quantity 100 g
Percent Purity 99.999%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'