Learn More
Iodic acid, 99.5%, for analysis, Thermo Scientific Chemicals
50.65 € - 572.85 €
Chemical Identifiers
CAS | 7782-68-5 |
---|---|
Molecular Formula | ILiO3 |
Molecular Weight (g/mol) | 181.84 |
MDL Number | MFCD00011348 |
InChI Key | FZAXZVHFYFGNBX-UHFFFAOYSA-M |
Synonym | unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min |
PubChem CID | 24345 |
ChEBI | CHEBI:24857 |
IUPAC Name | lithium(1+) iodate |
SMILES | [Li+].[O-][I](=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10606432
|
Thermo Scientific Acros
206450500 |
50 g | Glass bottle |
50.65 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10409910
|
Thermo Scientific Acros
206452500 |
250 g | Glass bottle |
163.82 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10590964
|
Thermo Scientific Acros
206450010 |
1 kg | Glass bottle |
572.85 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysisChemical Identifiers
7782-68-5 | |
181.84 | |
FZAXZVHFYFGNBX-UHFFFAOYSA-M | |
24345 | |
lithium(1+) iodate |
ILiO3 | |
MFCD00011348 | |
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min | |
CHEBI:24857 | |
[Li+].[O-][I](=O)=O |
Specifications
White to Beige | |
110.0°C | |
10ppm max. | |
99.5% | |
HIO3 | |
MFCD00011348 | |
unii-6u8j18jsbm, 6u8j18jsbm, iodicacid, monoiodic acid, caswell no. 499a, iodsaeure, trioxoiodic acid, iodic v acid, sodium iodate min | |
FZAXZVHFYFGNBX-UHFFFAOYSA-M | |
lithium(1+) iodate | |
24345 | |
175.91 | |
Analytical |
7782-68-5 | |
4.6200g/mL | |
4.62 | |
ILiO3 | |
Crystalline Powder | |
15, 5056 | |
Solubility in water: 2690g/l (20°C). Other solubilities: soluble in nitric acid, soluble in diluted alcohol, insoluble in absolute alcohol, ether and, chloroform | |
[Li+].[O-][I](=O)=O | |
181.84 | |
CHEBI:24857 | |
99.5% | |
Iodic acid, p.a. |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
missing translation for 'einecsNumber' : 231-962-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.