missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UltraPure™ 5M NaCl
Sterile-filtered, pre-mixed solution containing 292.2 g/L NaCl
Brand: Invitrogen™ 24740011
This item is not returnable.
View return policy
Description
- Common stock solution used in many molecular biology and forensic applications
- Performance and Quality Testing: No DNase, RNase, or protease activity detected
ChIP-on-Chip, Chromatin Biology, DNA & RNA Purification & Analysis, DNA Extraction, General gDNA Purification Reagents & Accessories, Genomic DNA Purification, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, RNAi, Epigenetics & Non-Coding RNA Research, Southern Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Sodium Chloride | |
Store at 15°C to 30°C. | |
UltraPure™ | |
10 L | |
Room Temperature |
5 M | |
Forensic, Molecular Biology | |
NaCl (Sodium Chloride) | |
Molecular Biology Grade | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.