Learn More
Invitrogen™ PureLink™ HiPure Plasmid Filter Midiprep Kit
PureLink HiPure Plasmid Filter Midiprep Kit is designed to isolate plasmid DNA of highest purity from 25 to 100 mL of overnight bacterial culture.
336.00 € - 592.00 €
Specifications
Column Type | Filter Column |
---|---|
Content And Storage | The PureLink™ HiPure Plasmid Filter Kits include Equilibration Buffer (EQ1), Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), RNase A, Wash Buffer (W8), Elution Buffer (E4), TE Buffer (TE), and HiPure Filter Columns. Store all components at room temperature upon receipt. |
Format | Column |
Isolation Technology | Anion Exchange Resin |
Sample Type | Bacterial Culture |
Includes: The PureLink HiPure Plasmid Filter Kits include Equilibration Buffer (EQ1), Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), RNase A, Wash Buffer (W8), Elution Buffer (E4), TE Buffer (TE), and HiPure Filter Columns.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10184912
|
Invitrogen™
K210014 |
25 Preps |
336.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10402522
|
Invitrogen™
K210015 |
50 Preps |
592.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PureLink HiPure Plasmid Filter Midiprep Kit is designed to isolate plasmid DNA of highest purity from 25 to 100 mL of overnight bacterial culture. The PureLink HiPure Plasmid Filter Kits include HiPure Filter Columns, which combine filtration for clarification of bacterial lysate with anion-exchange resin purification of plasmid DNA, in single unit. The resulting plasmid DNA is of quality equivalent to that obtained by purification with two passes through cesium chloride gradients—the most rigorous method for DNA purification.
- Purify high-quality DNA in Less than two hours for transfections and other applications with no need for additional steps to remove contaminants like RNA, proteins, or endotoxins
- Additionally, elimination of phenol, chloroform, ethidium bromide, and cesium chloride from protocol minimizes exposure to and disposal of hazardous materials
PureLink HiPure Plasmid Filter Kits provide:
- Simple protocols—bacterial lysate clarification without centrifugation
- Fast results save time by purifying plasmid DNA in less than two hours
- High yields up to 200 μg for midipreps and up to 850 μg for maxipreps
- Reliable performance, high-quality plasmid DNA purified with these kits is suitable for wide variety of applications
How it works
- Addition of a HiPure Filter Column to the PureLink HiPure Plasmid Kits provides rapid clearing of the bacterial lysate without the need for centrifugation
- Lysate filtration cartridge is integrated into DNA-binding column for a combined step of lysate clarification and plasmid DNA binding directly to the anion-exchange resin
- Single column wash removes impurities such as RNA, proteins, metabolites, and other low molecular weight molecules; the ultrapure plasmid DNA is then eluted with high-salt buffer
- DNA is then desalted and concentrated with an isopropanol precipitation step, then collected by centrifugation
Order Info
Shipping Condition: Room Temperature
Specifications
Filter Column | |
Column | |
Bacterial Culture | |
Plasmid DNA | |
Not High-throughput Compatible (Manual) | |
100-200 μg (Medium-Scale) Plasmid DNA | |
Plasmid Filter MidiPrep Kit | |
Room Temperature | |
PureLink™ | |
350 μg |
The PureLink™ HiPure Plasmid Filter Kits include Equilibration Buffer (EQ1), Resuspension Buffer (R3), Lysis Buffer (L7), Precipitation Buffer (N3), RNase A, Wash Buffer (W8), Elution Buffer (E4), TE Buffer (TE), and HiPure Filter Columns. Store all components at room temperature upon receipt. | |
Anion Exchange Resin | |
5 mL | |
Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription | |
<40kb, Low Copy Plasmid, High Copy Plasmid, BAC | |
PureLink™ | |
Transfection Grade | |
Plasmid DNA | |
90 min. |
For Research Use Only. Not for use in diagnostic procedures.