missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ Agarose Gels with SYBR™ Safe DNA Gel Stain, 1.2%
Less hazardous
Brand: Invitrogen™ G521801
Includes: Contains one multi-pack of 18 gels.
Description
- Ready for use with agarose, electrodes, and DNA stain packaged inside a dry, disposable, UV-transparent cassette
- There are no gels to pour, buffers to make, staining or destaining steps to perform, or gel boxes to assemble
- Offer excellent resolution and clarity in 12 minutes and are excellent for analyzing PCR products, restriction digests, and plasmid preparations
- 12-well, single comb gels
Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Contains one multi-pack of 18 gels. Store at room temperature. | |
E-Gel | |
12-well | |
SYBR Safe | |
Room Temperature |
1.2% | |
Agarose Gel | |
Less hazardous | |
E-Gel™ | |
Single Comb |
For Research Use Only. Not for use in diagnostic procedures.