missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BenchPro™ 4100 Reagent Vials
Compatible for use with the BenchPro™ 4100 Card Processing Station
Brand: Invitrogen™ WP3001
Includes: 50 graduated vials; Volume: 25mL
Description
Designed for use with WP0001.
Order Info
Shipping Condition: Room Temperature
Specifications
50 graduated vials; Volume: 25mLs (each). Store at room temperature. | |
Western Blot | |
BenchPro | |
Tube(s) | |
50 vials | |
50 tubes, 25 mL each |
25 mL | |
BenchPro 4100 Western Processing System | |
Reagent Vial | |
Designed Compatible Products WP0001 | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.