Learn More
INTS7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17054-1-AP-150UL
 View more versions of this product
Product Code. 16866874
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene encodes a subunit of the integrator complex. The integrator complex associates with the C-terminal domain of RNA polymerase II and mediates 3′-end processing of the small nuclear RNAs U1 and U2. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
Specifications
INTS7
Polyclonal
Unconjugated
INTS7
C1orf73, INT7, integrator complex subunit 7, INTS7
Rabbit
Antigen Affinity Chromatography
RUO
25896, 289382, 77065
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TQK1, Q9NVH2
Ints7
INTS7 Fusion Protein Ag10755
150 μL
Primary
Human, Mouse, Rat