Learn More
Astell Scientific™ Integral Steam Generator
Improve cycle times by increasing the efficiency of steam production. Astell Scientific™ Integral Steam Generator system also includes automatic water filling.
Specifications
For Use With (Equipment) | Autoclaves of 120 L Capacity and Above (Excluding The E-Range). |
---|---|
Product Type | Integral Steam Generator |
Includes | Automatic water filling |
Description
- Removes the heating elements from the chamber and replaces them with an integral generator for improved control over steam production.
- Autofill is also fitted as standard to the steam generator; eliminating the need to manually refill the chamber between cycles.
- A mains water supply enters the steam generator, where it is heated and converted to steam. This then enters the chamber directly from the steam generator as and when it is required.
Specifications
Autoclaves of 120 L Capacity and Above (Excluding The E-Range). | |
Integral Steam Generator | |
Automatic water filling |