Learn More
INSIG1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55282-1-AP-150UL
This item is not returnable.
View return policy
Description
The INSIG1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human INSIG1. This antibody recognizes human, mouse, rat antigen. The INSIG1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
INSIG1 | |
Polyclonal | |
Unconjugated | |
150 μL | |
3638 | |
IgG |
Western Blot, ELISA | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |