Learn More
Buerkle™ V4A Stainless Steel Inserts for Novartos Multi
The V4A Stainless Steel Inserts for Buerkle™ Novartos Multi fit precisely and can easily be replaced. Inserts are available in volumes from 0 to 2.0 ml.
Brand:  Buerkle™ 5358-0010
148.98 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11817281
  • 175.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Fit precisely
  • Can easily be replaced
  • Stainless steel AISI 316L
Specifications
Specifications
Insert
Autoclavable
FDA
Stainless Steel (V4A)
1 mL
Novartos Multi Sampler