missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ Mobile injector

Module E 944/2 for the optimum loading of narrow necked glassware 500 - 1,000 ml (PG 8527)

Brand:  Miele Professional™ 7459410

Product Code. 13208123

  • 1675.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

15 nozzles (E 352) 6 x 220 mm with clips (E 354)
Specifications

Specifications

Stainless Steel
Stainless Steel
Washer-disinfectors, laboratory
250 mm
Insert/Module for Narrow-necked Glassware
15 Narrow-necked glassware
352 mm
558 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers