missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20973-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody can recognize isoforms 1, 2, 4, and 5 of INSC.
In Drosophila, neuroblasts divide asymmetrically into another neuroblast at the apical side and a smaller ganglion mother cell on the basal side. Cell polarization is precisely regulated by 2 apically localized multiprotein signaling complexes that are tethered by Inscuteable, which regulates their apical localization.Specifications
INSC | |
Polyclonal | |
Unconjugated | |
INSC | |
INSC, Protein inscuteable homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
233752, 293166, 387755 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q1MX18, Q3HNM7 | |
INSC | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction