Learn More
Masterflex™ Inert in-line Check Valves
Designed for use with Cole-Parmer tubing. In-line check valves feature a smooth flow path that minimizes shear and turbulence.
Brand:  Masterflex™ 01355-00
Product Code. 11769609
  • 82.45 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Inert in-line Outlet Check Valve
  • No metal components allow inert flow path
  • No back flow leakage with back pressure of 0 to 100psi
  • Ideal for high-purity, low-pressure applications
  • Zero maintenance
Specifications
Specifications
EPDM, PEEK
1/4 in. to 28 UNF
100 psi
Inert in-line Outlet Check Valve
1/4 in. to 28 UNF