missing translation for 'onlineSavingsMsg'
Learn More

Indium tin oxide, Vacuum Deposition Grade, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11365407
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Indium tin oxide is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50926-11-9
Molecular Formula InOSn
Molecular Weight (g/mol) 249.527
MDL Number MFCD00171662
InChI Key AMGQUBHHOARCQH-UHFFFAOYSA-N
Synonym acmc-1au9m, tin ii oxide indigane
PubChem CID 57350325
IUPAC Name indium;oxotin
SMILES O=[Sn].[In]

Specifications

Physical Form Pieces
Quantity 5 g
Solubility Information Insoluble in water.
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Indium tin oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'