missing translation for 'onlineSavingsMsg'
Learn More

Indium tin oxide, 99.99% (metals basis), Thermo Scientific Chemicals

83.01 € - 729.63 €

Chemical Identifiers

CAS 50926-11-9
Molecular Formula InOSn
Molecular Weight (g/mol) 249.527
MDL Number MFCD00171662
InChI Key AMGQUBHHOARCQH-UHFFFAOYSA-N
Synonym acmc-1au9m, tin ii oxide indigane
PubChem CID 57350325
IUPAC Name indium;oxotin
SMILES O=[Sn].[In]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11353438
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036348.06
5 g
83.01 €
5g
Please to purchase this item. Need a web account? Register with us today!
11363438
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036348.14
25 g
216.08 €
25g
Please to purchase this item. Need a web account? Register with us today!
11343438
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036348.22
100 g
729.63 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indium tin oxide is used in nanotechnology to provide a path to a new generation of solar cells, to make transparent conductive coatings for displays such as liquid crystal displays, flat panel displays, plasma displays, touch panels, and electronic ink. Thin films of ITO are also used in organic light-emitting diodes, antistatic coatings and EMI shielding. In organic light-emitting diodes, ITO is used as the anode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50926-11-9
249.527
AMGQUBHHOARCQH-UHFFFAOYSA-N
57350325
O=[Sn].[In]
InOSn
MFCD00171662
acmc-1au9m, tin ii oxide indigane
indium;oxotin

Specifications

50926-11-9
Powder
InOSn
acmc-1au9m, tin ii oxide indigane
AMGQUBHHOARCQH-UHFFFAOYSA-N
indium;oxotin
57350325
White powder
White
99.99% (Metal basis)
MFCD00171662
Insoluble in water.
O=[Sn].[In]
249.527
Odorless
Indium tin oxide
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P271-P281-P304+P340-P308+P313-P501c

H335-H351

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only