missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) perchlorate octahydrate, 99.9% (metals basis), Thermo Scientific™

Product Code. 11381748
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-15-1
MDL Number MFCD00150241

Specifications

Melting Point 800°C
Physical Form Crystalline
Quantity 50 g
UN Number UN1481
Solubility Information It is soluble in water.
Formula Weight 557.30 (413.18 Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Indium(III) perchlorate octahydrate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.