missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indium(III) oxide, 99.994% (metals basis), Thermo Scientific Chemicals
120.00 € - 1350.00 €
Chemical Identifiers
CAS | 1312-43-2 |
---|---|
Molecular Formula | In2O3 |
Molecular Weight (g/mol) | 277.63 |
MDL Number | MFCD00011060 |
InChI Key | PJXISJQVUVHSOJ-UHFFFAOYSA-N |
IUPAC Name | diindium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[In+3].[In+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11351918
|
Thermo Scientific Alfa Aesar
011853.09 |
10 g |
120.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11361918
|
Thermo Scientific Alfa Aesar
011853.18 |
50 g |
375.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11341918
|
Thermo Scientific Alfa Aesar
011853.30 |
250 g |
1350.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Indium(III) oxide is used as a chemical reagent and intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1312-43-2 | |
277.63 | |
PJXISJQVUVHSOJ-UHFFFAOYSA-N | |
[O--].[O--].[O--].[In+3].[In+3] |
In2O3 | |
MFCD00011060 | |
diindium(3+) trioxidandiide |
Specifications
850° volat. | |
Yellow | |
99.994% (Metals basis) | |
MFCD00011060 | |
PJXISJQVUVHSOJ-UHFFFAOYSA-N | |
diindium(3+) trioxidandiide | |
277.64 | |
7.18 |
1312-43-2 | |
Powder | |
In2O3 | |
Insoluble in water. | |
[O--].[O--].[O--].[In+3].[In+3] | |
277.63 | |
Odorless | |
Indium(III) oxide |
Safety and Handling
missing translation for 'einecsNumber' : 215-193-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only