Learn More
Indium(III) hydroxide, 99.998% (metals basis), Thermo Scientific Chemicals
Description
Indium(III) hydroxide, acts as a precursor to indium(III) oxide. It is used in batteries, glasses, ceramics, chemical reagents, semiconductor materials. It is also used used for manufacturing of panel displays and demist.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 150°C (in -H2O) |
Physical Form | Powder |
Quantity | 50 g |
Solubility Information | It is insoluble in water. |
Formula Weight | 165.84 |
Percent Purity | 99.998% |
Assay | (metals basis) |
Chemical Name or Material | Indium(III) hydroxide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.