missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) chloride hydrate, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11371928
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Indium(III) chloride hydrate is used for mild and selective cleavage of tert-butyldimethylsilyl ethers. It is a Lewis acid, and serves as a catalyst for many organic transformations. It also catalyzes the conversion of aldehydes to gem diacetates by chemoselective process. Lithium indium hydride, an in situ reducing agent, can be prepared from indium trichloride. It catalyzes reactions such as Friedel-Crafts acylations and Diels-Alder reactions. It also catalyzes Knoevenagel condensation between the barbituric acid and the aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143983-91-9
Molecular Formula Cl3In
Molecular Weight (g/mol) 221.17
MDL Number MFCD00011058
InChI Key PSCMQHVBLHHWTO-UHFFFAOYSA-K
IUPAC Name indium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[In+3]

Specifications

Color White
Physical Form Crystalline
Quantity 250 g
UN Number UN3260
Merck Index 14,4958
Solubility Information Soluble in water,diethyl ether,and THF.
Formula Weight 221.18 (Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Indium(III) chloride hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'