missing translation for 'onlineSavingsMsg'
Learn More

Indium(III) acetate, 99.99% (metals basis), Thermo Scientific Chemicals

90.05 € - 302.51 €

Chemical Identifiers

CAS 25114-58-3
Molecular Formula C2H4InO2
Molecular Weight (g/mol) 174.87
MDL Number MFCD00054347
InChI Key LGHOYKSQIQISBI-UHFFFAOYSA-N
Synonym indium acetate, acetic acid, indium 3+ salt
PubChem CID 24191761
IUPAC Name acetic acid;indium
SMILES CC(=O)O.[In]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369877
SDS View Documents
Thermo Scientific Alfa Aesar
042230.09
10 g
90.05 €
10g
Please to purchase this item. Need a web account? Register with us today!
11379877
SDS View Documents
Thermo Scientific Alfa Aesar
042230.18
50 g
302.51 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Indium(III) acetate is used as a catalyst in intermolecular radical addition of organic iodides to electron deficient alkenes. It is also used in the reduction of 1-bromo-3-phenylpropane to propylbenzene. Further, it is used to prepare the indium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25114-58-3
174.87
LGHOYKSQIQISBI-UHFFFAOYSA-N
24191761
CC(=O)O.[In]
C2H4InO2
MFCD00054347
indium acetate, acetic acid, indium 3+ salt
acetic acid;indium

Specifications

25114-58-3
99.99% (metals basis)
MFCD00054347
Soluble in water,mineral acid and acetic acid.
CC(=O)O.[In]
174.87
291.96
Indium(III) acetate
Powder
C2H4InO2
indium acetate, acetic acid, indium 3+ salt
LGHOYKSQIQISBI-UHFFFAOYSA-N
acetic acid;indium
24191761
Hygroscopic
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Indium(III) acetate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Acetic acid, indium(3+) salt

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.