Learn More
Thermo Scientific Chemicals Indigo, synthetic
47.03 € - 122.61 €
Chemical Identifiers
CAS | 482-89-3 |
---|---|
Molecular Formula | C16H10N2O2 |
Molecular Weight (g/mol) | 262.268 |
MDL Number | MFCD00005722 |
InChI Key | COHYTHOBJLSHDF-BUHFOSPRSA-N |
Synonym | C.I. 73000, Vat Blue 1 |
PubChem CID | 5318432 |
IUPAC Name | (2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one |
SMILES | C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10206940
|
Thermo Scientific Chemicals
212130250 |
25 g | Glass bottle |
47.03 €
25g |
In Stock
Log in to see stock. |
||||
10082650
|
Thermo Scientific Chemicals
212131000 |
100 g | Glass bottle |
122.61 €
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
482-89-3 | |
262.268 | |
COHYTHOBJLSHDF-BUHFOSPRSA-N | |
5318432 | |
C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2 |
C16H10N2O2 | |
MFCD00005722 | |
C.I. 73000, Vat Blue 1 | |
(2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one |
Specifications
482-89-3 | |
MFCD00005722 | |
15, 4981 | |
COHYTHOBJLSHDF-BUHFOSPRSA-N | |
C1=CC=C2C(=C1)C(=O)C(=C3C(=O)C4=CC=CC=C4N3)N2 | |
262.268 | |
262.26 | |
Blue to Violet | |
Indigo, synthetic |
C16H10N2O2 | |
24, 417 | |
C.I. 73000, Vat Blue 1 | |
Authentic | |
(2E)-2-(3-oxo-1H-indol-2-ylidene)-1H-indol-3-one | |
5318432 | |
Complies | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 207-586-9
missing translation for 'rtecsNumber' : DU2988400
missing translation for 'tsca' : TSCA
- Indigo
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.