missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Indigo Carmine, High Purity Biological Stain
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
Dye Content | 85% min. |
Beilstein | 25, 304 |
Merck Index | 15, 4982 |
Solubility Information | Solubility in water: 1g/100mL (25 c). Other solubilities: slightly soluble in alcohol, practically insoluble in most other organic, solvents |
Infrared Spectrum | Authentic |
Formula Weight | 466.34 |
Grade | Analysis |
Packaging | Glass bottle |
Color | Blue |
Quantity | 100 g |
Show More |
Safety and Handling
Hazard Category
- Skin sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction