Learn More
QIAGEN Indicating FTA™ Micro Card
Collect, stabilize, process, transport, and archive buccal DNA samples. Indicating FTA™ Micro Cards are available as indicating cards for use with clear samples, such as buccal cells or saliva.
Brand:  QIAGEN WB120211
Product Code. 11320854
  • 600.00 € / Pack of 100
Explore more special offers
Description
Description
  • Improves process efficiency by enabling direct amplification for STR analysis due to immediate DNA capture on the FTA card.
  • Includes an inert dye, which turns to white when a buccal sample or saliva is added for easy confirmation of sample addition.
  • Immediate stabilization and protection of nucleic acids for long term archiving of samples at room temperature.
  • Different card options to support manual or automated processing for versatility and incorporation into current workflows.
Specifications
Specifications
Card
For ensics, Transgenic Identification, Transfusion Medicine/HLA Typing
1
125 μL
DNA
Inhert
Indicating FTA Micro Card
125μL
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
QIAGEN Indicating FTA™ Micro Card > For Use With: Forensics, Transgenic Identification, Transfusion Medicine/HLA Typing