Learn More
IL5RA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12655-1-AP-150UL
 View more versions of this product
Product Code. 16800494
  • 410.00 € / 150µL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Interleukin 5 (IL-5) is a soluble T cell-derived factor, also known as T cellreplacing factor (TRF), that induces B cell and eosinophil growth and differentiation. IL-5 exerts its biological activity through the IL-5 receptor (IL-5R),which is composed of two chains: an alpha chain that binds IL-5 with low affinity and a beta chain that does not bind IL-5, but together with the IL-5 alpha chain,constitutes the high affinity IL-5 receptor. The cytoplasmic domain of both the alpha and beta chains is essential for signal transduction. Specifically, the membrane-proximal proline-rich sequence of the ytoplasmic domain of the IL-5R receptor alpha subunit, IL-5Ralpha, is critical for the IL-5 induced proliferative response, expression of nuclear proto-oncogenes and tyrosine phosphorylation of cellular proteins, such as JAK1 and JAK2.
Specifications
Specifications
IL5RA
Polyclonal
Unconjugated
IL5RA
CD125, CDw125, HSIL5R3, IL 5 receptor subunit alpha, IL 5R alpha, IL 5R subunit alpha, IL 5RA, IL5R, IL5RA, interleukin 5 receptor, alpha
Rabbit
Antigen Affinity Chromatography
RUO
114103, 16192, 3568
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21183, Q01344
Il5ra
IL5RA Fusion Protein Ag3358
150 μL
Primary
Human, Mouse, Rat