missing translation for 'onlineSavingsMsg'
Learn More

IFI27 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA555997

Product Code. 15753911

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: MEASALTSSA VTSVAKVVRV ASGSAVVLPL ARIATV Highest antigen sequence identity to the following orthologs: Mouse - 39%, Rat - 39%.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IFI27
Polyclonal
Unconjugated
IFI27
IFI27
Recombit protein corresponding to Human IFI27
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P40305
Rabbit
Antigen affinity chromatography
RUO
3429
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only