Learn More
Manganese Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand:  SPEX Certiprep CLMS-1
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 10318633
  • 319.00 € / 125mL
Explore more special offers
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Ce10μg/mL
Dy10μg/mL
Er10μg/mL
Eu10μg/mL
Gd10μg/mL
Ho10μg/mL
La10μg/mL
Lu10μg/mL
Nd10μg/mL
Pr10μg/mL
Sc10μg/mL
Sm10μg/mL
Tb10μg/mL
Th10μg/mL
Tm10μg/mL
Y10μg/mL
Yb10μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Multi-Element Solution 1
125 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Each component at 10μg/mL in 5% HNO3:
Cerium
Dysprosium
Erbium
Europium
Gadolinium
Holmium
Lanthanum
Lutetium
Neodymium
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Trace ICP/ICP-MS
5% HNO3
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product intended for laboratory use only