missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12759-1-AP-20UL
This item is not returnable.
View return policy
Description
DNA-dependent RNA polymerase catalyzes the transcription of DNA into RNA using the four ribonucleoside triphosphates as substrates. Component of RNA polymerase I which synthesizes ribosomal RNA precursors. Appears to be involved in the formation of the initiation complex at the promoter by mediating the interaction between Pol I and UBTF/UBF.Specifications
ICAM5 | |
Polyclonal | |
Unconjugated | |
ICAM5 | |
ICAM 5, ICAM5, Telencephalin, TLCN, TLN | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15898, 313785, 7087 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q60625, Q9UMF0 | |
Icam5 | |
ICAM5 Fusion Protein Ag3303 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction