missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ICAM-1 Mouse ELISA Kit

Sandwich ELISA Kit

612.00 € - 4815.00 €

Specifications

Accession Number P13597
Assay Sensitivity 20 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15405283
View Documents Promotion Details
Invitrogen™
EMICAM1ALPHA
96 Tests
612.00 €
1 set
Estimated Shipment: 22-11-2024
to see stock.
Add to basket
 
17115739
View Documents Promotion Details
Invitrogen™
EMICAM1ALPHAX5
5 x 96 Tests
2635.00 €
Each
Estimated Shipment: 06-12-2024
to see stock.
Add to basket
 
18587874
View Documents Promotion Details
Invitrogen™
EMICAM1ALPHAX10
10 x 96 Tests
4815.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Mouse Intercellular Adhesion Molecule 1 (Ms ICAM-1) ELISA quantitates Ms ICAM-1 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms ICAM-1. Principle of the method The Mouse ICAM-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

ICAM-1 (CD54) is an 85-110 kDa single-chain type 1 integral membrane glycoprotein with an extracellular domain of five immunoglobulin superfamily repeats, a transmembrane region and a cytoplasmic domain. ICAM-1 has 7 potential N-linked glycosylation sites and shares considerable amino acid sequence homology with ICAM-3 (CD50) and ICAM-2 (CD102). ICAM-1 binds to integrins of type CD11a/CD18 (leukocyte adhesion molecule, LFA-1), or CD11b/CD18 (Mac-1) and is exploited by Rhinovirus as a receptor. ICAM-1 is expressed by activated endothelial cells and detected on epithelial cells, fibroblasts, chondrocytes, B lymphocytes, T lymphocytes (low), monocytes, macrophages, dendritic cells and neutrophils, with lower levels that increase upon inflammation. ICAM-1 is also detected in some carcinoma and melanoma cells. Soluble ICAM-1 is detectable in the plasma and is elevated in patients with various inflammatory syndromes.
Specifications

Specifications

P13597
Solid-phase sandwich ELISA
ELISA Kit
Mouse
Colorimetric Microplate Reader
CD54,Icam-1,Ly-47,MALA-2
<10%
RUO
2°C to 8°C
Mouse
4 hr. 45 min.
20 pg/mL
Biotin
Other Proteins
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
15894
<12%
HRP
Inflammation
CD54, Icam-1, Ly-47, MALA-2
1 hr. 20 min.
Videos
SDS
Documents

Documents

Product Certifications