Learn More
Invitrogen™ mirVana™ miRNA Mimic, miR-1 Positive Control
miRNA mimics are small, chemically modified double-stranded RNA molecules designed to specifically bind to and mimic endogenous miRNA molecules and enable miRNA functional analysis by up-regulation of miRNA activity.
Brand: Invitrogen™ 4464063
817.40 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
miRNA mimics are small, chemically modified double-stranded RNA molecules designed to specifically bind to and mimic endogenous miRNA molecules and enable miRNA functional analysis by up-regulation of miRNA activity.
We have now released the second generation of miRNA mimics, mirVana™ miRNA Mimics, which:
- Exhibit maximum and consistent effect in vitro at low concentration
- Offer superior specificity due to unique Star strand modification
- Target the most updated Sanger content
- Can be used in vitro and in vivo, offering you consistency throughout your entire research project
- Are non-toxic
Positive and negative controls are also available for use along with mirVana™ miRNA Mimics in miRNA up-regulation experiments.
mirVana™ miRNA Mimic miR-1 Positive Control
The mirVana™ miRNA Mimic miR-1 Positive Control is designed to mimic mature miR-1 miRNA after delivery to mammalian cells. miR-1 is associated with down-regulation of many genes. In particular, miR-1-mediated down-regulation of Protein Tyrosine Kinase 9 (PTK9) has been shown to occur at the mRNA level. miR-1 Positive Control is intended for use as a positive control for transfection experiments using mirVana™ miRNA Mimics. Monitor activity of miR-1 Positive Control through its gene silencing of PTK9, using real-time reverse transcription-PCR to detect PTK9 mRNA.
First Generation Mimic Controls
Our first-generation miRNA mimics (and controls), Ambion™ Pre-miR™ miRNA Precursors, have lower specificity and potency, plus a lower strand specificity compared to the new mirVana™ miRNA Mimics. These first-generation mimics are recommended for completing ongoing in vitro miRNA studies. For new studies or in vivo studies, we highly recommend our new mirVana™ miRNA Mimics and their corresponding controls.
Specifications
Store at or below -20°C. Do not store in a frost-free freezer. | |
Positive Control | |
miRNA & Non-Coding RNA Analysis | |
mirVana™ | |
Purified | |
Room Temperature |
mirVana™ miRNA Mimic, 20nmol, Liquid, Unconjugated Dye, Pre-miRNA RNAi Type, Positive Control Type, Used for miRNA and Non-Coding RNA Analysis, Purified Purification, –20°C Storage Temperature, For Research Use Only | |
miRNA Mimic | |
Unconjugated | |
miRNA Mimic | |
20 nmol |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.