Learn More
Thermo Scientific™ Hypersil™ PREP HS C8 HPLC Columns
Use these preparative columns for scale-up purification with high phase loading. They are suitable for all USP-L7 applications.
149.54 € - 16943.09 €
Specifications
pH | 2 to 8 |
---|---|
Pore Size | 100 Å |
Surface Area | 350 m²/g |
Temperature | 60°C |
USP Type | L7 |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Column Format | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Particle Size | Column Format | Price | Quantity & Availability | ||||||
16428473
|
Thermo Scientific™
41103-052130 |
50 mm | 2 mm | 3 μm | Preparative Column, Standard |
497.25 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16438473
|
Thermo Scientific™
41103-054630 |
50 mm | 4.6 mm | 3 μm | Preparative Column, Standard |
552.65 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16448473
|
Thermo Scientific™
41103-102130 |
100 mm | 2 mm | 3 μm | Preparative Column, Standard |
622.80 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16458473
|
Thermo Scientific™
41103-104630 |
100 mm | 4.6 mm | 3 μm | Preparative Column, Standard |
678.19 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16468473
|
Thermo Scientific™
41103-152130 |
150 mm | 2 mm | 3 μm | Preparative Column, Standard |
691.73 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16478473
|
Thermo Scientific™
41103-154630 |
150 mm | 4.6 mm | 3 μm | Preparative Column, Standard |
761.89 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
16488473
|
Thermo Scientific™
41103-252130 |
250 mm | 2 mm | 3 μm | Preparative Column, Standard |
807.43 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16498473
|
Thermo Scientific™
41103-254630 |
250 mm | 4.6 mm | 3 μm | Preparative Column, Standard |
836.96 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16408483
|
Thermo Scientific™
41105-052130 |
50 mm | 2 mm | 5 μm | Preparative Column, Standard |
427.10 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16418483
|
Thermo Scientific™
41105-054630 |
50 mm | 4.6 mm | 5 μm | Preparative Column, Standard |
494.80 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16428483
|
Thermo Scientific™
41105-059070A |
50 mm | 10 mm | 5 μm | Preparative Column, Standard |
1165.60 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16438483
|
Thermo Scientific™
41105-059270A |
50 mm | 20 mm | 5 μm | Preparative Column, Standard |
1234.52 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16448483
|
Thermo Scientific™
41105-059270XL |
50 mm | 25 mm | 5 μm | Preparative Column, Xtended Life |
4623.81 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16458483
|
Thermo Scientific™
41105-102130 |
100 mm | 2 mm | 5 μm | Preparative Column, Standard |
552.65 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16468483
|
Thermo Scientific™
41105-104630 |
100 mm | 4.6 mm | 5 μm | Preparative Column, Standard |
609.27 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16478483
|
Thermo Scientific™
41105-109070A |
100 mm | 10 mm | 5 μm | Preparative Column, Standard |
1234.52 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16488483
|
Thermo Scientific™
41105-109270A |
100 mm | 20 mm | 5 μm | Preparative Column, Standard |
2329.96 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16498483
|
Thermo Scientific™
41105-109270XL |
100 mm | 25 mm | 5 μm | Preparative Column, Xtended Life |
6147.30 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16408493
|
Thermo Scientific™
41105-152130 |
150 mm | 2 mm | 5 μm | Preparative Column, Standard |
599.41 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16418493
|
Thermo Scientific™
41105-154630 |
150 mm | 4.6 mm | 5 μm | Preparative Column, Standard |
678.19 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16428493
|
Thermo Scientific™
41105-159070A |
150 mm | 10 mm | 5 μm | Preparative Column, Standard |
1411.76 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16438493
|
Thermo Scientific™
41105-159270A |
150 mm | 20 mm | 5 μm | Preparative Column, Standard |
2786.60 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16448493
|
Thermo Scientific™
41105-159270XL |
150 mm | 25 mm | 5 μm | Preparative Column, Xtended Life |
6717.63 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16458493
|
Thermo Scientific™
41105-159370A |
150 mm | 30 mm | 5 μm | Preparative Column, Standard |
4555.29 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16468493
|
Thermo Scientific™
41105-159370XL |
150 mm | 30 mm | 5 μm | Preparative Column, Xtended Life |
8319.88 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16478493
|
Thermo Scientific™
41105-159570A |
150 mm | 50 mm | 5 μm | Preparative Column, Standard |
9083.52 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16488493
|
Thermo Scientific™
41105-159570XL |
150 mm | 50 mm | 5 μm | Preparative Column, Xtended Life |
14335.03 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16498493
|
Thermo Scientific™
41105-252130 |
250 mm | 2 mm | 5 μm | Preparative Column, Standard |
702.81 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16408503
|
Thermo Scientific™
41105-254630 |
250 mm | 4.6 mm | 5 μm | Preparative Column, Standard |
807.43 €
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
16418503
|
Thermo Scientific™
41105-259070A |
250 mm | 10 mm | 5 μm | Preparative Column, Standard |
1607.14 €
Each |
Estimated Shipment: 26-05-2025
Log in to see stock. |
||||||
16428503
|
Thermo Scientific™
41105-259270A |
250 mm | 20 mm | 5 μm | Preparative Column, Standard |
3352.78 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16438503
|
Thermo Scientific™
41105-259270XL |
250 mm | 25 mm | 5 μm | Preparative Column, Xtended Life |
6576.50 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16448503
|
Thermo Scientific™
41105-259370A |
250 mm | 30 mm | 5 μm | Preparative Column, Standard |
5408.94 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16458503
|
Thermo Scientific™
41105-259370XL |
250 mm | 30 mm | 5 μm | Preparative Column, Xtended Life |
8788.90 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16468503
|
Thermo Scientific™
41105-259570A |
250 mm | 50 mm | 5 μm | Preparative Column, Standard |
10219.58 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16478503
|
Thermo Scientific™
41105-259570XL |
250 mm | 50 mm | 5 μm | Preparative Column, Xtended Life |
16943.09 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16488503
|
Thermo Scientific™
41110-018823A |
8 mm | 10 mm | 10 μm | Guard Column |
149.54 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16498503
|
Thermo Scientific™
41110-019023A |
20 mm | 10 mm | 10 μm | Guard Column |
261.70 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16408513
|
Thermo Scientific™
41110-019323A |
30 mm | 10 mm | 10 μm | Guard 1/16 in. |
317.13 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16418513
|
Thermo Scientific™
41110-019323B |
30 mm | 10 mm | 10 μm | Guard 1/8 in. |
317.13 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16428513
|
Thermo Scientific™
41110-029023A |
20 mm | 20 mm | 10 μm | Guard Column | N/A | N/A | N/A | |||||
16438513
|
Thermo Scientific™
41110-029323A |
30 mm | 20 mm | 10 μm | Guard 1/16 in. |
523.40 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16448513
|
Thermo Scientific™
41110-029323B |
30 mm | 20 mm | 10 μm | Guard 1/8 in. |
523.40 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16458513
|
Thermo Scientific™
41110-059070A |
50 mm | 10 mm | 10 μm | Preparative Column, Standard |
1095.44 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16468513
|
Thermo Scientific™
41110-059270A |
50 mm | 20 mm | 10 μm | Preparative Column, Standard |
1128.68 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16478513
|
Thermo Scientific™
41110-059270XL |
50 mm | 25 mm | 10 μm | Preparative Column, Xtended Life |
4564.11 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16488513
|
Thermo Scientific™
41110-109070A |
100 mm | 10 mm | 10 μm | Preparative Column, Standard |
1165.60 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16498513
|
Thermo Scientific™
41110-109270A |
100 mm | 20 mm | 10 μm | Preparative Column, Standard |
1400.69 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16408523
|
Thermo Scientific™
41110-109270XL |
100 mm | 25 mm | 10 μm | Preparative Column, Xtended Life |
5080.12 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16418523
|
Thermo Scientific™
41110-159070A |
150 mm | 10 mm | 10 μm | Preparative Column, Standard |
1341.60 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16428523
|
Thermo Scientific™
41110-159270A |
150 mm | 20 mm | 10 μm | Preparative Column, Standard |
2716.44 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16438523
|
Thermo Scientific™
41110-159270XL |
150 mm | 25 mm | 10 μm | Preparative Column, Xtended Life |
5761.99 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16448523
|
Thermo Scientific™
41110-159370A |
150 mm | 30 mm | 10 μm | Preparative Column, Standard |
4963.95 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16458523
|
Thermo Scientific™
41110-159370XL |
150 mm | 30 mm | 10 μm | Preparative Column, Xtended Life |
8521.26 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16468523
|
Thermo Scientific™
41110-159570A |
150 mm | 50 mm | 10 μm | Preparative Column, Standard |
9083.52 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16478523
|
Thermo Scientific™
41110-159570XL |
150 mm | 50 mm | 10 μm | Preparative Column, Xtended Life |
13088.51 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16488523
|
Thermo Scientific™
41110-259070A |
250 mm | 10 mm | 10 μm | Preparative Column, Standard |
1534.49 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16498523
|
Thermo Scientific™
41110-259270A |
250 mm | 20 mm | 10 μm | Preparative Column, Standard |
3246.92 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16408533
|
Thermo Scientific™
41110-259270XL |
250 mm | 25 mm | 10 μm | Preparative Column, Xtended Life |
6066.58 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16418533
|
Thermo Scientific™
41110-259370A |
250 mm | 30 mm | 10 μm | Preparative Column, Standard |
5031.70 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16428533
|
Thermo Scientific™
41110-259370XL |
250 mm | 30 mm | 10 μm | Preparative Column, Xtended Life |
8528.90 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16438533
|
Thermo Scientific™
41110-259570A |
250 mm | 50 mm | 10 μm | Preparative Column, Standard |
9935.27 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
16448533
|
Thermo Scientific™
41110-259570XL |
250 mm | 50 mm | 10 μm | Preparative Column, Xtended Life |
13945.97 €
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
Description
Get maximum loading for your scale up from analytical to a prep LC method with Thermo Scientific™ Hypersil™ PREP HS C8 reversed phase preparative HPLC column. With similar selectivity to C18 columns and predictable elution order, C8 columns are ideal for analytes with medium hydrophobicity or when a less hydrophobic phase is required to obtain optimum retention. The lower hydrophobicity allows compounds to elute more quickly, yielding faster separations. By providing high surface area particles with a 100Å pore size, and a variety of dimensions including an option for XtendedLife hardware, which improves column lifetime by maintaining a uniform packed bed.
Similar selectivity, less retention than C18 columns
- Successful retention and separation of moderately hydrophobic analytes.
- Ideal for fatty acids, which are too hydrophobic for analysis using C18 columns.
- C8 (USP L7) selectivity - high surface area particle with 100Å pore size.
- pH stable from 2 to 8.
- Maximum temperature of 60°C.
Faster separations
- Higher analytical throughput compared to C18 alkyl chain chemistry.
- Quicker compound elution due to reduced hydrophobic interactions.
Preparative HPLC Column Hardware Options - based on column inner diameter (i.d.) and column performance needs.
Standard Threaded and Standard Flange Hardware
- Designed for standard applications, expertly packed and robustly designed to provide versatile products for routine use.
- Standard threaded hardware for column i.d. < 40mm.
- Standard flange hardware for column i.d. ≥ 40mm.
- Standard hardware can be modified on request to be compatible with SFC.
XtendedLife Hardware
- Improves column lifetime by incorporating an internal Dynamic Axial Compression (DAC) system, ensuring the media bed is continuously packed tightly.
- Compatible with SFC and is available in dimensions up to 50mm i.d.
Preparative Guards are a proven and cost effective way to extend the lifetime of your prep column. Our preparative guard columns require the use of a preparative guard holder.
Customer Specific Orders
- If there is a Thermo Scientific stationary phase you are currently using and would like to obtain in a preparative scale not shown or would like to discuss a specific column dimension, we would be happy to discuss your requirements.
- Please contact your local sales representative for Customer Specific Orders.
- Custom built columns are not eligible for returns.
Specifications
2 to 8 | |
350 m²/g | |
L7 | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase |
100 Å | |
60°C | |
C8 | |
7% | |
Yes |