Learn More
Thermo Scientific™ Hypersil™ BDS C8 Javelin™ Guard Columns
Offer convenient, easy-to-use protection for analytical HPLC columns
375.00 €
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) | Price | Quantity & Availability | ||||
11573161
|
Thermo Scientific™
28205-022106 |
2.1 mm | 20 mm | N/A | |||||
Description
- Direct-connection, fingertight design for convenience and maximum efficiency
- Patented design requires no holder, allowing quick and easy replacement
Features of BDS C8 Solid Phase
- Excellent choice for acids, bases and neutral compounds
- Less retentive than C18 for faster analyses
- Robust columns with long column lifetimes
- Very reliable for routine assays
Specifications
5 μm | |
C8 |