missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-917 Partial Nitrile-Coated Seamless Knit Liquid Repellant Gloves
GREENER_CHOICE

Product Code. 16029681
Click to view available options
Size:
6
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves repel oil, grease, and dirt making them ideal for use in grimey work environments. Seamless nylon liner provides comfort, dexterity, and a secure fit.

  • Liquid repellent against oil, grease, and dirt
  • Seamless nylon liner
  • Suitable for food handling
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Nitrile
Color Blue
Hand Specific
Material Nylon
Size 6
EN388 Standard 4121A
For Use With (Application) Maintenance, Quality Control, and Metal Fabrication
Glove Cuff Color White
Glove Cuff Style Knit
Series 11-917
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.