missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-739 Polyurethane-Coated Extended Cuff Gloves with Intercept™ Technology

Product Code. 16639093
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves combine comfort and dexterity with Intercept™ Technology to provide a high level of cut resistance. Polyurethane coating enhances grip capabilities in dry, as well as slightly oily environments.

  • A combination of comfort, sure handling, and cut resistance
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Ideal for safe use in controlling, and handling/assembly applications that involve contact with sharp pieces
  • Designed for use in dry as well as slightly oily environments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE 0493, EN 407:2004, EN 420:2003 + A1:2009
Coating Material Polyurethane
Color Gray
Hand Specific
Material Intercept Technology Yarn
Size 8
EN388 Standard 4X43D
For Use With (Application) Assembly, Metal Parts Handling, and Press Injection Molding
Glove Cuff Style Knit
Series 11-739
Yarn Gauge 13
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'