missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-728 Polyurethane-Coated HPPE-Lined Gloves with Intercept™ Technology

Product Code. 16649083
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves combine comfort and dexterity with Intercept™ Technology to provide a high level of cut resistance. Polyurethane coating enhances grip capabilities in dry, as well as slightly oily environments.

  • A combination of comfort, sure handling, and cut resistance
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Ideal for safe use in controlling, and handling/assembly applications that involve contact with sharp pieces
  • Designed for use in dry as well as slightly oily environments
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Gray
Hand Specific
Material Intercept Technology Yarn
Size 11
EN388 Standard 4342B
For Use With (Application) Glass Repair, Metal Handling, and Press Injection Molding
Glove Cuff Style Knit
Liner Material HPPE
Series 11-728
Yarn Gauge 15
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'