missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-651 Medium Duty Cut-Resistant Industrial Gloves

Product Code. 16049641
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Intercept™ Cut Resistance Technology blends engineered and synthetic fibers into high performance yarns to provide cut protection, along with comfort and dexterity.

  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Constructed with a seamless, 13-gauge liner
  • Elastic knit wrist ensures comfort, dexterity and a secure fit
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 6
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Cuff Material Elastic
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A3
Hand Specific
Material Intercept Technology Yarn
Size 10
EN388 Standard 4X42C
For Use With (Application) Machinery, Equipment Operation, and Metal Fabrication
Glove Cuff Style Knit
Series 11-651
Yarn Gauge 13
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'