missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-515 Hi-Vis Cut-Resistant Nitrile Gloves with Intercept Technology™ Construction

Product Code. 16059601
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Intercept Technology™ yarn incorporates DuPont™ Aramid Fiber Stretch Armor for ANSI Level 5 Cut Performance. Gloves are ergonomically designed with enhanced grip capabilities in dry or light oil applications.

  • Gloves combine comfort and durability with ANSI Level 5 Cut Performance
  • Ergonomically designed with hi-vis colors to better protect users in low light environments
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Extended cuff provides additional cut protection in the highly sensitive wrist area
  • Foam nitrile formulation provides longer wear for increased durability
  • Enhances grip and tactility in light oil or dry environments for grip assurance when handling sharp objects and knives
  • Provides workers with the ability to perform safely and efficiently
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Coating Material Nitrile
Color Orange
Cut Resistance Rating ANSI A5
Hand Specific
Material Intercept Technology Yarn
Size 11
EN388 Standard 4X42E
For Use With (Application) Automotive Assembly, Metal Fabrication
Gender Unisex
Glove Cuff Style Knit
Series 11-515
Yarn Gauge 13
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'