missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-515 Hi-Vis Cut-Resistant Nitrile Gloves with Intercept Technology™ Construction

Product Code. 16009601
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Intercept Technology™ yarn incorporates DuPont™ Aramid Fiber Stretch Armor for ANSI Level 5 Cut Performance. Gloves are ergonomically designed with enhanced grip capabilities in dry or light oil applications.

  • Gloves combine comfort and durability with ANSI Level 5 Cut Performance
  • Ergonomically designed with hi-vis colors to better protect users in low light environments
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Extended cuff provides additional cut protection in the highly sensitive wrist area
  • Foam nitrile formulation provides longer wear for increased durability
  • Enhances grip and tactility in light oil or dry environments for grip assurance when handling sharp objects and knives
  • Provides workers with the ability to perform safely and efficiently
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Coating Material Nitrile
Color Orange
Cut Resistance Rating ANSI A5
Hand Specific
Material Intercept Technology Yarn
Size 6
EN388 Standard 4X42E
For Use With (Application) Automotive Assembly, Metal Fabrication
Gender Unisex
Glove Cuff Style Knit
Series 11-515
Yarn Gauge 13
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'