missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11488356
Click to view available options
Quantity:
100 g
250 g
500 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxylamine hydrochloride is a monomoamine oxidase inhibitor. It is used to prepare oximes and hydroxmic acids in organic synthesis. It acts as a copolymerization inhibitor. It can be used to remove bromine and polybromide from a solution during extraction of lignin from lignocellulosic biomass. It is key starting material for the preparation of pharmaceuticals and agrochemicals. It plays a vital role in rubber and plastic industries as an antioxidant, a vulcanization accelerator and a radical scavenger. It is also used as a color stabilizer and emulsion additive in color films.

Solubility
Soluble in water, alcohol, methanol and glycerol.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, phosphorous pentachloride, calcium and anhydrous copper(II) sulfate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine;hydrochloride
SMILES Cl.NO

Specifications

Melting Point 152°C (dec.)
pH 3.6
Quantity 5000 g
Linear Formula NH2OH·HCl
UN Number UN2923
Beilstein 3539763
Merck Index 14,4828
Solubility Information Soluble in water,alcohol,methanol and glycerol.
Formula Weight 69.49
Percent Purity 99%
Odor Acidic
Sensitivity Hygroscopic
Density 1.67
Chemical Name or Material Hydroxylamine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'