Learn More
Poulten Selfe Hydrometer petroleum density 0.950 to 1.000g/mL graduated for measurements at 15°C Poulten Selfe and Lee
Brand:  Poulten Selfe 430/08
Hydrometer petroleum density 0.950 to 1.000g/mL graduated for measurements at 15°C Poulten Selfe and Lee
Code : 02
Additional Details : Weight : 1.00000kg
Product Code. 12931448
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.