Learn More
hydrogen tetrachloroaurate(III) hydrate, Thermo Scientific Chemicals
195.00 € - 785.00 €
Chemical Identifiers
CAS | 27988-77-8 |
---|---|
Molecular Formula | AuCl4H |
Molecular Weight (g/mol) | 339.77 |
MDL Number | MFCD00149903 |
InChI Key | RFWCEGCWRSNAGT-UHFFFAOYSA-K |
Synonym | Chloroauric acid hydrate, Gold trichloride acid hydrate |
IUPAC Name | hydrogen tetrachlorogoldtris(ylium) |
SMILES | [H+].Cl[Au+3](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10791111
|
Thermo Scientific Acros
223620010 |
1 g | Glass bottle |
195.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001611
|
Thermo Scientific Acros
223620050 |
5 g | Glass bottle |
785.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27988-77-8 | |
339.77 | |
RFWCEGCWRSNAGT-UHFFFAOYSA-K | |
hydrogen tetrachlorogoldtris(ylium) |
AuCl4H | |
MFCD00149903 | |
Chloroauric acid hydrate, Gold trichloride acid hydrate | |
[H+].Cl[Au+3](Cl)(Cl)Cl |
Specifications
30.0°C | |
Orange | |
AuCl4H | |
MFCD00149903 | |
Chloroauric acid hydrate, Gold trichloride acid hydrate | |
RFWCEGCWRSNAGT-UHFFFAOYSA-K | |
hydrogen tetrachlorogoldtris(ylium) | |
339.78 | |
Hydrogen tetrachloroaurate(III) hydrate, p.a. |
27988-77-8 | |
Crystalline Powder and/or Crystals | |
HAuCl4·xH2O | |
14, 4522 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether | |
[H+].Cl[Au+3](Cl)(Cl)Cl | |
339.77 | |
≥49% (Au) |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
GHS Signal Word: Danger
- Hydrogen tetrachloroaurate(III) hydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only